Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011787.hg.1 |
ENST00000527364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011787.hg.1 |
ENST00000527364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011787.hg.1 |
ENST00000527364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011787.hg.1 |
ENST00000527364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038296.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010569.hg.1 |
NR_038296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000425384.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
ENST00000435043.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010569.hg.1 |
NR_038296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011787.hg.1 |
ENST00000527364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011787.hg.1 |
ENST00000527364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011787.hg.1 |
ENST00000527364.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011787.hg.1 |
ENST00000527364.1 |
Ensembl |