Assay Details
Gene Symbol: LINC00680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK023611.1 | - | 1 | 325 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000399751.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000418765.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000422882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000423899.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | ENST00000450081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_003660.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125727.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125728.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014303.hg.1 | NR_125729.1 | Refseq |
