Assay Details
| Gene Symbol: POM121C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127026.1 | - | 1 | 513 | 79 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000415277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000415277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000415277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000415277.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000614583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000614583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000614583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000614583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000398379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000415277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000415277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000415277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000415277.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000439629.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000473609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000479864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000607367.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000614583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000614583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000614583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000614583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | ENST00000615331.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011550.hg.1 | NM_001099415.2 | Refseq |

POM121C
Genomic Map