Assay Details
| Gene Symbol: MRPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011520276.2 | XP_011518578.1 | 6 | 2102 | 77 | |
| GenBank mRNA | AL833465.1 | - | 1 | 2410 | 77 | |
More Information
Set Membership: |
|

MRPL23
Genomic Map