Assay Details
Gene Symbol: TSPAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK055832.1 | - | 2 | 822 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571707.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000571914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000574882.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000611590.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | ENST00000621293.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_001290212.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NM_031945.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009184.hg.1 | NR_110866.1 | Refseq |
