Assay Details
| Gene Symbol: WEE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015392.1 | 7 | 1931 | 77 | ||
| GenBank mRNA | AK098171.1 | - | 7 | 1931 | 77 | |
| BC101358.2 | - | 4 | 1375 | 77 | ||
| BC101359.2 | - | 7 | 2185 | 77 | ||
| BC101360.2 | - | 3 | 1221 | 77 | ||
| BC131564.1 | - | 5 | 1561 | 77 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000473776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000473776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000459753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000462383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000465110.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000471512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000473776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000473776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000478332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000484172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000486906.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000488785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | ENST00000495800.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012839.hg.1 | NR_015392.1 | Refseq |

WEE2-AS1
Genomic Map