Assay Details
| Gene Symbol: EXTL3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC043205.2 | - | 2 | 1557 | 111 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000519870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000519870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000519870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000519870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000523789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000523789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000523789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | ENST00000523789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000501224.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000519870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000519870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000519870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000519870.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000520023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000523789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000523789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000523789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | ENST00000523789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | NR_126027.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009987.hg.1 | NR_126027.1 | Refseq |

EXTL3-AS1
Genomic Map