Assay Details
Gene Symbol: LRRC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321963.1 | NP_001308892.1 | 1 | 37 | 77 | |
NM_001321966.1 | NP_001308895.1 | 1 | 80 | 77 | ||
NR_135909.1 | 1 | 80 | 77 | |||
NR_135910.1 | 1 | 37 | 77 | |||
XM_017013297.1 | XP_016868786.1 | 1 | 80 | 77 | ||
GenBank mRNA | DA359044.1 | - | 1 | 80 | 77 | |
DA789153.1 | - | 1 | 114 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001321963.1, NM_001321966.1, XM_017013297.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
