Assay Details
Gene Symbol: LRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR749434.1 | - | 2 | 4283 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000276654.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000424843.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000518375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000519675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000520770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000522046.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | ENST00000523007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_001135703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011395.hg.1 | NM_013437.4 | Refseq |
