Assay Details
Gene Symbol: CYP11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017013146.1 | XP_016868635.1 | 1 | 221 | 62 | |
GenBank mRNA | AK094090.1 | - | 1 | 325 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK094090.1, XM_017013146.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
C21-steroid hormone biosynthetic process
glucocorticoid biosynthetic process
immune response
cholesterol metabolic process
regulation of blood pressure
sterol metabolic process
aldosterone biosynthetic process
cellular response to hormone stimulus
cortisol biosynthetic process
cellular response to potassium ion
glucose homeostasis
secondary metabolite biosynthetic process
oxidation-reduction process
cellular response to peptide hormone stimulus
steroid 11-beta-monooxygenase activity
iron ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
heme binding
corticosterone 18-monooxygenase activity
C21-steroid hormone biosynthetic process
glucocorticoid biosynthetic process
immune response
cholesterol metabolic process
regulation of blood pressure
sterol metabolic process
aldosterone biosynthetic process
cellular response to hormone stimulus
cortisol biosynthetic process
cellular response to potassium ion
glucose homeostasis
secondary metabolite biosynthetic process
oxidation-reduction process
cellular response to peptide hormone stimulus
steroid 11-beta-monooxygenase activity
iron ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
heme binding
corticosterone 18-monooxygenase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000292427.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000314111.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000377675.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000517471.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | ENST00000519285.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_000497.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012092.hg.1 | NM_001026213.1 | Refseq |
