Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000379142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
ENST00000379142.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000297625.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000379142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
ENST00000379142.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009894.hg.1 |
NM_020702.4 |
Refseq |