Assay Details
| Gene Symbol: C9orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096971.1 | - | 1 | 1020 | 81 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000495934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000495934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000495934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000372447.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461539.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000461762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000480023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000492991.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000495934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000495934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000495934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | ENST00000615599.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011710.hg.1 | NM_016520.2 | Refseq |

C9orf78
Genomic Map