Assay Details
| Gene Symbol: FAM120AOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322224.1 | NP_001309153.1 | 1 | 204 | 132 | |
| NR_136232.1 | 1 | 204 | 132 | |||
| NR_136233.1 | 1 | 204 | 132 | |||
| NR_136234.1 | 1 | 204 | 132 | |||
| NR_136235.1 | 1 | 204 | 132 | |||
| NR_136236.1 | 1 | 204 | 132 | |||
| NR_136237.1 | 1 | 204 | 132 | |||
| NR_136238.1 | 1 | 204 | 132 | |||
| GenBank mRNA | BC063650.1 | - | 1 | 97 | 132 | |
| BC080634.1 | - | 1 | 74 | 132 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001322224.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000428378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000375412.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000423591.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000428378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000476484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000479094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000483149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000520470.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | ENST00000523407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010837.hg.1 | NM_198841.3 | Refseq |

FAM120AOS
Genomic Map