Assay Details
| Gene Symbol: LINC00671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027254.1 | 4 | 792 | 60 | ||
| GenBank mRNA | AK055784.1 | - | 4 | 902 | 60 | |
| AK298293.1 | - | 3 | 723 | 60 | ||
| BC122867.1 | - | 3 | 622 | 60 | ||
| BC122868.1 | - | 4 | 714 | 60 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010034.hg.1 | ENST00000408294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000301683.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000417193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000431109.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | ENST00000436546.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010751.hg.1 | NR_027254.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010034.hg.1 | ENST00000408294.1 | Ensembl |

LINC00671
Genomic Map