Assay Details
Gene Symbol: LOC730098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320038.1 | NP_001306967.1 | 1 | 192 | 68 | |
GenBank mRNA | AK296005.1 | - | 1 | 152 | 68 | |
BC066358.1 | - | 1 | 185 | 68 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK125920.1 | - | 1 | Chr. 9 - 34665496 - 34669257 [+] on Build GRCh38 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK296005.1, NM_001320038.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900012121.hg.1 | ENST00000434627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012121.hg.1 | ENST00000434627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012121.hg.1 | ENST00000434627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012121.hg.1 | ENST00000434627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012121.hg.1 | ENST00000434627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012121.hg.1 | ENST00000434627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
