Assay Details
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318025.1 | NP_001304954.1 | 8 | 2641 | 110 | |
NM_001318026.1 | NP_001304955.1 | 9 | 2583 | 110 | ||
NM_001318027.1 | NP_001304956.1 | 8 | 2579 | 110 | ||
NM_174907.3 | NP_777567.1 | 9 | 2812 | 110 | ||
GenBank mRNA | AL833216.1 | - | 6 | 2246 | 110 | |
BC110889.1 | - | 8 | 2610 | 6503859 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00008930.hg.1 | ENST00000445107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008930.hg.1 | ENST00000445107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008930.hg.1 | ENST00000445107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008930.hg.1 | ENST00000445107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000356692.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000460360.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000470976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000476505.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000482242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000488810.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | ENST00000495566.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013847.hg.1 | NM_174907.3 | Refseq |
