Assay Details
Gene Symbol: CHST11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173982.1 | NP_001167453.1 | 1 | 309 | 109 | |
NM_018413.5 | NP_060883.1 | 1 | 309 | 109 | ||
GenBank mRNA | AB042326.1 | - | 1 | 197 | 109 | |
AK290923.1 | - | 1 | 250 | 109 | ||
AK313217.1 | - | 1 | 252 | 109 | ||
DA853534.1 | - | 1 | 310 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB042326.1, AK290923.1, NM_001173982.1, NM_018413.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chondrocyte development
respiratory gaseous exchange
post-embryonic development
carbohydrate biosynthetic process
chondroitin sulfate biosynthetic process
negative regulation of transforming growth factor beta receptor signaling pathway
polysaccharide localization
post-anal tail morphogenesis
regulation of cell proliferation
embryonic digit morphogenesis
negative regulation of apoptotic process
developmental growth
embryonic viscerocranium morphogenesis
N-acetylgalactosamine 4-O-sulfotransferase activity
chondroitin 4-sulfotransferase activity
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase activity
chondrocyte development
respiratory gaseous exchange
post-embryonic development
carbohydrate biosynthetic process
chondroitin sulfate biosynthetic process
negative regulation of transforming growth factor beta receptor signaling pathway
polysaccharide localization
post-anal tail morphogenesis
regulation of cell proliferation
embryonic digit morphogenesis
negative regulation of apoptotic process
developmental growth
embryonic viscerocranium morphogenesis
N-acetylgalactosamine 4-O-sulfotransferase activity
chondroitin 4-sulfotransferase activity
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000550711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | ENST00000550711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000303694.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000546689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000547956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000549260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000550711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | ENST00000550711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_001173982.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008683.hg.1 | NM_018413.5 | Refseq |
