| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000470091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000470091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000470091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000470091.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000477702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000477702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000477702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000480237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000480237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000480237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000489452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
ENST00000489452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
NR_027345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
NR_027345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
NR_027345.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
NR_027346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
NR_027346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
NR_027346.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012711.hg.1 |
NR_027346.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000470091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000470091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000470091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000470091.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000477702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000477702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000477702.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000480237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000480237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000480237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000489452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
ENST00000489452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
NR_027345.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
NR_027345.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
NR_027345.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
NR_027346.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
NR_027346.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
NR_027346.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012711.hg.1 |
NR_027346.1 |
Refseq |