Assay Details
Gene Symbol: HLA-G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF196178.1 | - | 4 | 923 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000360323.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376815.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376818.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000376828.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000428701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478355.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | ENST00000478519.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007487.hg.1 | NM_002127.5 | Refseq |
