Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011860.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011860.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011860.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011860.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011860.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011860.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011860.hg.1 |
ENST00000478808.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011860.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011860.hg.1 |
NR_038996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011860.hg.1 |
NR_038996.1 |
Refseq |