Assay Details
Gene Symbol: LOC399815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027282.1 | 5 | 73 | |||
GenBank mRNA | AK300776.1 | - | 5 | 73 | ||
DY654994.1 | - | 2 | 204 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000368895.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | ENST00000425266.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009163.hg.1 | NR_027282.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000368891.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000462191.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000470158.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | ENST00000481909.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012088.hg.1 | NM_024942.3 | Refseq |
