Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009095.hg.1 |
ENST00000423739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009095.hg.1 |
ENST00000423739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009095.hg.1 |
ENST00000423739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009095.hg.1 |
ENST00000423739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009095.hg.1 |
NR_047022.1 |
Refseq |