Assay Details
| Gene Symbol: NRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB593101.1 | - | 1 | 122 | 97 | |
| AB593103.1 | - | 1 | 122 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB593101.1, AB593103.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |

NRL
Genomic Map