Assay Details
| Gene Symbol: C14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008404.2 | NP_001008404.1 | 5 | 1276 | 102 | |
| NM_001286399.1 | NP_001273328.1 | 5 | 1232 | 102 | ||
| NM_001286400.1 | NP_001273329.1 | 5 | 1441 | 102 | ||
| XM_005267638.3 | XP_005267695.1 | 5 | 1345 | 102 | ||
| XM_011536764.2 | XP_011535066.1 | 5 | 1323 | 102 | ||
| GenBank mRNA | AK095913.1 | - | 5 | 1241 | 102 | |
| AK297818.1 | - | 5 | 1143 | 102 | ||
| BC041103.1 | - | 5 | 1178 | 5193008 | 102 | |
| BX161457.1 | - | 5 | 1441 | 102 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010368.hg.1 | ENST00000556073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010368.hg.1 | ENST00000556073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010368.hg.1 | ENST00000556073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010368.hg.1 | ENST00000556073.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000331952.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000410013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | ENST00000557649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001008404.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008443.hg.1 | NM_001286400.1 | Refseq |

C14orf180
Genomic Map