Assay Details
| Gene Symbol: TMC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120365.1 | 3 | 517 | 99 | ||
| GenBank mRNA | BI821075.1 | - | 3 | 432 | 99 | |
| CB134362.1 | - | 3 | 434 | 99 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | ENST00000560973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559277.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559277.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000559781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | ENST00000560973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008062.hg.1 | NR_120365.1 | Refseq |

TMC3-AS1
Genomic Map