| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010534.hg.1 |
ENST00000420691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010534.hg.1 |
ENST00000420691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010534.hg.1 |
ENST00000420691.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010534.hg.1 |
NR_024160.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010534.hg.1 |
NR_024160.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010534.hg.1 |
NR_024160.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010534.hg.1 |
ENST00000420691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010534.hg.1 |
ENST00000420691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010534.hg.1 |
ENST00000420691.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010534.hg.1 |
NR_024160.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010534.hg.1 |
NR_024160.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010534.hg.1 |
NR_024160.1 |
Refseq |