Assay Details
| Gene Symbol: RHOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004040.3 | NP_004031.1 | 1 | 1402 | 86 | |
| GenBank mRNA | AK124398.1 | - | 1 | 1399 | 86 | |
| AK309991.1 | - | 2 | 1015 | 86 | ||
| AK310813.1 | - | 2 | 769 | 86 | ||
| BC066954.1 | - | 1 | 1399 | 5264057 | 86 | |
More Information
Set Membership: |
|

RHOB
Genomic Map