Assay Details
Gene Symbol: ZNF880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145434.1 | NP_001138906.1 | 4 | 1956 | 174 | |
GenBank mRNA | AF086165.1 | - | 1 | 228 | 174 | |
AK001357.1 | - | 1988 | 174 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
