Assay Details
| Gene Symbol: LINC00671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027254.1 | 4 | 1410 | 78 | ||
| GenBank mRNA | AK055784.1 | - | 4 | 1520 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00671| Gene Symbol: LINC00671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027254.1 | 4 | 1410 | 78 | ||
| GenBank mRNA | AK055784.1 | - | 4 | 1520 | 78 | |
Set Membership: |
|