Assay Details
Gene Symbol: LOC101928304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125855.1 | 4 | 604 | 131 | ||
NR_125856.1 | 3 | 488 | 131 | |||
GenBank mRNA | CA430304.1 | - | 1 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000418837.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000423028.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000606039.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | ENST00000607531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014342.hg.1 | NR_125856.1 | Refseq |
