Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000439703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013996.hg.1 |
ENST00000449466.1 |
Ensembl |