Assay Details
| Gene Symbol: CBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127228.1 | NP_001120700.1 | 5 | 1523 | 95 | |
| NM_006807.4 | NP_006798.1 | 5 | 1713 | 95 | ||
| GenBank mRNA | BC002609.1 | - | 5 | 1447 | 3140815 | 95 |
| BC021302.2 | - | 5 | 1514 | 5013102 | 95 | |
| U35451.1 | - | 5 | 1448 | 95 | ||
More Information
Set Membership: |
|

CBX1
Genomic Map