Assay Details
| Gene Symbol: CARD8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040599.1 | 2 | 1727 | 137 | ||
| GenBank mRNA | AK125548.1 | - | 2 | 1723 | 137 | |
| BC060888.1 | - | 2 | 1712 | 137 | ||
More Information
Set Membership: |
|

CARD8-AS1
Genomic Map