Assay Details
| Gene Symbol: TNKS2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108070.1 | 2 | 577 | 77 | ||
| NR_108071.1 | 2 | 248 | 77 | |||
| GenBank mRNA | AA776692.1 | - | 1 | 77 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | ENST00000432246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | ENST00000432246.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | ENST00000432938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | ENST00000432938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | ENST00000432938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | NR_108070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | NR_108070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | NR_108070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | NR_108071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | NR_108071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011414.hg.1 | NR_108071.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | ENST00000432246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | ENST00000432246.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | ENST00000432938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | ENST00000432938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | ENST00000432938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | NR_108070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | NR_108070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | NR_108070.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | NR_108071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | NR_108071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011414.hg.1 | NR_108071.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000432541.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000433969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000434993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000442719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000446784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | ENST00000456577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_002331.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109980.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012363.hg.1 | NR_109981.1 | Refseq |

TNKS2-AS1
Genomic Map