Assay Details
Gene Symbol: TIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003255.4 | NP_003246.1 | 5 | 815 | 62 | |
GenBank mRNA | AK294290.1 | - | 5 | 575 | 62 | |
AL713764.1 | - | 4 | 2156 | 62 | ||
BC039613.1 | - | 4 | 553 | 62 | ||
BC040039.1 | - | 4 | 465 | 62 | ||
BC040445.1 | - | 4 | 550 | 62 | ||
BC052605.1 | - | 5 | 792 | 6267022 | 62 | |
BC071586.1 | - | 5 | 812 | 30337524 | 62 | |
J05593.1 | - | 5 | 783 | 62 | ||
M32304.1 | - | 5 | 767 | 62 | ||
S48568.1 | - | 5 | 783 | 62 | ||
X54533.1 | - | 5 | 426 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
central nervous system development
aging
negative regulation of cell proliferation
response to hormone
negative regulation of endopeptidase activity
extracellular matrix disassembly
regulation of Rap protein signal transduction
response to cytokine
response to drug
positive regulation of MAPK cascade
positive regulation of neuron differentiation
positive regulation of adenylate cyclase activity
negative regulation of mitotic cell cycle
negative regulation of Ras protein signal transduction
negative regulation of membrane protein ectodomain proteolysis
protease binding
integrin binding
protein binding
metalloendopeptidase inhibitor activity
metal ion binding
central nervous system development
aging
negative regulation of cell proliferation
response to hormone
negative regulation of endopeptidase activity
extracellular matrix disassembly
regulation of Rap protein signal transduction
response to cytokine
response to drug
positive regulation of MAPK cascade
positive regulation of neuron differentiation
positive regulation of adenylate cyclase activity
negative regulation of mitotic cell cycle
negative regulation of Ras protein signal transduction
negative regulation of membrane protein ectodomain proteolysis
protease binding
integrin binding
protein binding
metalloendopeptidase inhibitor activity
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000619342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000619342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000619342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | ENST00000619342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000262768.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000322630.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000536189.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000585421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586057.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000586713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000587052.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000590267.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000592761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000619342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000619342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000619342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | ENST00000619342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243540.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_001243541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011919.hg.1 | NM_003255.4 | Refseq |
