Assay Details
Gene Symbol: NPTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX537550.1 | - | 1 | 2720 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000306773.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000535681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000571100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | ENST00000575212.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011984.hg.1 | NM_002522.3 | Refseq |
