| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |