Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000321751.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338338.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000338370.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000378853.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000489799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
ENST00000496905.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127229.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_001127230.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012471.hg.1 |
NM_017900.2 |
Refseq |