Assay Details
| Gene Symbol: SH3D21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162530.1 | NP_001156002.1 | 14 | 1400 | 80 | |
| NM_024676.4 | NP_078952.4 | 11 | 1206 | 80 | ||
| XM_011542153.2 | XP_011540455.1 | 14 | 1445 | 80 | ||
| XM_017002340.1 | XP_016857829.1 | 14 | 1445 | 80 | ||
| XM_017002341.1 | XP_016857830.1 | 14 | 1445 | 80 | ||
| XM_017002342.1 | XP_016857831.1 | 14 | 1445 | 80 | ||
| XM_017002343.1 | XP_016857832.1 | 14 | 1445 | 80 | ||
| XM_017002344.1 | XP_016857833.1 | 14 | 1445 | 80 | ||
| XM_017002345.1 | XP_016857834.1 | 14 | 1445 | 80 | ||
| XM_017002346.1 | XP_016857835.1 | 6 | 817 | 80 | ||
| GenBank mRNA | AK056459.1 | - | 7 | 901 | 80 | |
| AK297015.1 | - | 11 | 1203 | 80 | ||
| BC101676.1 | - | 3 | 447 | 80 | ||
| BC101678.1 | - | 2 | 394 | 80 | ||
| BG003040.1 | - | 1 | 80 | |||
More Information
Set Membership: |
|

SH3D21
Genomic Map