Assay Details
Gene Symbol: RGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK093544.1 | - | 3 | 290 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000462589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000462589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000462589.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000474373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000474373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000474373.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000583155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000583155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000583155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | ENST00000583155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000367459.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000462589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000462589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000462589.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000469578.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000474373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000474373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000474373.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000498352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000583155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000583155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000583155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | ENST00000583155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | NM_002922.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011012.hg.1 | NM_002922.3 | Refseq |
