Assay Details
Gene Symbol: LOC102724264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134483.1 | 2 | 97 | |||
NR_134484.1 | 2 | 97 | ||||
GenBank mRNA | AA868454.1 | - | 1 | 153 | 97 | |
AI685301.1 | - | 1 | 167 | 97 | ||
AI914404.1 | - | 1 | 136 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
