| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000453816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000453816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000441223.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000453816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000453816.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
ENST00000543490.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010580.hg.1 |
NM_001102659.2 |
Refseq |