Assay Details
Gene Symbol: AGPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011512043.2 | XP_011510345.1 | 2 | 218 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011512043.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000460342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000460342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | ENST00000460342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000264167.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000409888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000460342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000460342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | ENST00000460342.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010100.hg.1 | NM_003659.3 | Refseq |
