Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037631.2 | NP_001032720.1 | 2 | 380 | 119 | |
| NM_005214.4 | NP_005205.2 | 2 | 380 | 119 | ||
| GenBank mRNA | AF414120.1 | - | 2 | 380 | 119 | |
| AF486806.1 | - | 2 | 225 | 119 | ||
| AK313732.1 | - | 2 | 365 | 119 | ||
| AY209009.1 | - | 2 | 225 | 119 | ||
| BC069566.1 | - | 2 | 320 | 7262243 | 119 | |
| BC070162.1 | - | 2 | 381 | 30417685 | 119 | |
| BC074842.2 | - | 2 | 251 | 30915552 | 119 | |
| BC074893.2 | - | 2 | 256 | 30915247 | 119 | |
| CD639535.1 | - | 2 | 384 | 119 | ||
| L15006.1 | - | 2 | 225 | 119 | ||
| U90273.1 | - | 1 | 114 | 119 | ||
More Information
Set Membership: |
|

CTLA4
Genomic Map