Assay Details
Gene Symbol: C1orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011544124.2 | XP_011542426.1 | 6 | 1874 | 104 | |
XM_011544125.2 | XP_011542427.1 | 5 | 1624 | 104 | ||
XM_011544128.2 | XP_011542430.1 | 5 | 2135 | 104 | ||
XM_011544129.2 | XP_011542431.1 | 6 | 1785 | 104 | ||
XM_011544130.2 | XP_011542432.1 | 5 | 1706 | 104 | ||
XM_017000571.1 | XP_016856060.1 | 6 | 1904 | 104 | ||
XM_017000572.1 | XP_016856061.1 | 6 | 2412 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
