Assay Details
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003262.3 | NP_003253.1 | 8 | 1660 | 168 | |
XM_011513114.2 | XP_011511416.1 | 9 | 1737 | 168 | ||
GenBank mRNA | AK294504.1 | - | 3 | 824 | 168 | |
BX648539.1 | - | 8 | 1644 | 168 | ||
D87127.1 | - | 8 | 1641 | 168 | ||
U93239.1 | - | 8 | 1641 | 168 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000337002.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000460513.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000461933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469515.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000469890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000470355.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000480708.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000481435.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000487736.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | ENST00000497277.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009458.hg.1 | NM_003262.3 | Refseq |
