Assay Details
| Gene Symbol: SEC16A-NOTCH1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SEC16A-NOTCH1_S1N27.0 | - | 57 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC16A-NOTCH1 fusion| Gene Symbol: SEC16A-NOTCH1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SEC16A-NOTCH1_S1N27.0 | - | 57 | 84 | ||