Assay Details
| Gene Symbol: VAMP2-NRG1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_VAMP2-NRG1_V4N4.0 | - | 117 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VAMP2-NRG1 fusion| Gene Symbol: VAMP2-NRG1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_VAMP2-NRG1_V4N4.0 | - | 117 | 93 | ||