Assay Details
| Gene Symbol: BRCA2-BRCA2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BRCA2-BRCA2_B11B27_V12.0 | - | 68 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRCA2-BRCA2 fusion| Gene Symbol: BRCA2-BRCA2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_BRCA2-BRCA2_B11B27_V12.0 | - | 68 | 99 | ||