Assay Details
| Gene Symbol: FOXP1-PDGFRA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FOXP1-PDGFRA_F16P12.0 | - | 44 | 117 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXP1-PDGFRA fusion| Gene Symbol: FOXP1-PDGFRA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FOXP1-PDGFRA_F16P12.0 | - | 44 | 117 | ||
Set Membership: |
|